accessibility menu, dialog, popup

UserName

CAS RN 16903-61-0

BH4CuPPPhPhPhPhPhPh

CAS RN 16903-61-0

IUPAC Name: bis(triphenylphosphane) copper boranuide
Synonyms: bis(triphenylphosphine) copper boranuide
Molecular Weight (g/mol): 602.97
Molecular Formula: C36H34BCuP2
InChi Key: OIFZQKCJHVNBRA-UHFFFAOYSA-N
SMILES: [BH4-].[Cu].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results