accessibility menu, dialog, popup

UserName

CAS RN 169126-63-0

BCH3H3CH3CCH3OHHO

CAS RN 169126-63-0

IUPAC Name: (5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)boronic acid
Synonyms: 5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-ylboronic acid
Molecular Weight (g/mol): 232.13
Molecular Formula: C14H21BO2
InChi Key: NXBNRLONOXGRCQ-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results