CAS RN 169332-60-9
IUPAC Name:
(4S)-4-[(2S)-3-carboxy-2-acetamidopropanamido]-4-{[(1S)-1-{[(2S)-1-carboxy-3-oxopropan-2-yl]carbamoyl}-2-methylpropyl]carbamoyl}butanoic acid
Synonyms:
ac-asp-glu-val-asp-H
Molecular Weight (g/mol):
502.48
Molecular Formula:
C20H30N4O11
InChi Key:
UMBVAPCONCILTL-MRHIQRDNSA-N
SMILES:
CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C=O
1
–
3
of
3
results
1
MedChemExpress Ac-DEVD-CHO
MedChemExpress Ac-DEVD-CHO is a specific Caspase-3 inhibitor with a Ki value of 230 pM.
2
MedChemExpress Ac-DEVD-CHO
MedChemExpress Ac-DEVD-CHO is a specific Caspase-3 inhibitor with a Ki value of 230 pM.
3
Thermo Scientific™ N-Acetyl-Asp-Glu-Val-Asp-al
CAS: 169332-60-9 Molecular Formula: C20H30N4O11 Synonym: Ac-Asp-Glu-Val-Asp-CHO; Ac-DEVD-CHO