missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Asp-Glu-Val-Asp-al

Product Code. 15416789
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416789 1 mg 1mg
15406789 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416789 Supplier Thermo Scientific Chemicals Supplier No. J65204.M
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169332-60-9
Molecular Formula C20H30N4O11
Synonym Ac-Asp-Glu-Val-Asp-CHO; Ac-DEVD-CHO

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Reversible inhibitor of interleukin-1beta converting enzyme (ICE); also inhibits caspase-3 cleavage of poly(ADP-ribose) polymerase
Formula Weight 502.47
Sensitivity Moisture Sensitive
Product Type N-Acetyl-Asp-Glu-Val-Asp-al
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.