accessibility menu, dialog, popup

UserName

FFFFFFSbH

CAS RN 16950-06-4

IUPAC Name: fluoroantimonic acid
Synonyms: fluoroantimonic acid
Molecular Weight (g/mol): 236.76
Molecular Formula: F6HSb
InChi Key: OFBVOLRJPPBHCL-UHFFFAOYSA-O
SMILES: [H+].[F].[F].[F].[F].[F].[F].[Sb]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)