missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexafluoroantimonate(V), ≈65% aq. soln., Thermo Scientific Chemicals

Product Code. 11377738
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377738 25 g 25g
11367738 50 g 50g
11347738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377738 Supplier Thermo Scientific Alfa Aesar Supplier No. 033488.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16950-06-4

Hydrogen hexafluoroantimonate(V) is an excellent medium for the preparation of alkylidene oxonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16950-06-4
MDL Number MFCD00011327
Synonym Hexafluoroantimonic acid; Fluoroantimonic acid

Specifications

Color Colorless to Orange
Linear Formula HF·SbF5
Quantity 25 g
UN Number UN3264
Solubility Information It is fully miscible with water,soluble in SO2ClF,SO2.
Formula Weight 236.75
Concentration or Composition (by Analyte or Components) ≈65% aq. soln.
Physical Form Liquid
Chemical Name or Material Hydrogen hexafluoroantimonate(V)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.