accessibility menu, dialog, popup

UserName

CAS RN 169566-81-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2,3-dimethylbutanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: AWEZXIRZNQCCNN-UHFFFAOYNA-N
SMILES: CC(C)C(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)