CAS RN 169675-09-6
CAS RN 169675-09-6
IUPAC Name:
(2E)-but-2-enedioic acid; (2S)-1-(6-chloro-5-fluoro-1H-indol-1-yl)propan-2-amine
Synonyms:
(2S)-1-(6-chloro-5-fluoroindol-1-yl)propan-2-amine; fumaric acid
Molecular Weight (g/mol):
342.75
Molecular Formula:
C15H16ClFN2O4
InChi Key:
CEPHEXXZGQBXGT-XZTIXWOLSA-N
SMILES:
OC(=O)\C=C\C(O)=O.C[C@H](N)CN1C=CC2=CC(F)=C(Cl)C=C12
1
–
1
of
1
results
1
Tocris Bioscience™ Ro 60-0175 fumarate
CAS: 169675-09-6 Molecular Formula: C15H16ClFN2O4