missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ro 60-0175 fumarate
GREENER_CHOICE

Product Code. 30001311
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001311

Brand: Tocris Bioscience™ 1854/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective 5-HT2C agonist

Potent, selective 5-HT2 receptor agonist; shows selectivity for the 5-HT2C subtype (pKi values are 9, 7.5, 5.4, 5.2 and 5.6 for human 5-HT2C, 2A, 1A, 6 and 7 receptors respectively). Centrally active following oral or systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169675-09-6
Molecular Formula C15H16ClFN2O4

Specifications

Quantity 50 mg
Formula Weight 342.75
Purity >98%
Molecular Weight (g/mol) 342.75
Chemical Name or Material Ro 60-0175 fumarate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.