CAS RN 170569-86-5
CAS RN 170569-86-5
IUPAC Name:
4-[5-(4-chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzene-1-sulfonamide
Synonyms:
4-[5-(4-chlorophenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
Molecular Weight (g/mol):
401.79
Molecular Formula:
C16H11ClF3N3O2S
InChi Key:
NSQNZEUFHPTJME-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=C(C=C1)N1N=C(C=C1C1=CC=C(Cl)C=C1)C(F)(F)F