missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SC 236

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective COX-2 inhibitor (IC50 values are 0.005 and 17.8 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory properties and potent antimetastatic activity against both experimental metastases and spontaneous metastases arising following primary tumor excision.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 401.79 |
| Purity | >99% |
| Molecular Weight (g/mol) | 401.788 |
| Chemical Name or Material | SC 236 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction