accessibility menu, dialog, popup

UserName

HOOFF

CAS RN 170924-50-2

IUPAC Name: [4-(difluoromethoxy)phenyl]methanol
Synonyms: [4-(difluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: HSIDWXYUJAUALR-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results