accessibility menu, dialog, popup

UserName

CAS RN 1719-57-9

CH3SiCH3ClCl

CAS RN 1719-57-9

IUPAC Name: chloro(chloromethyl)dimethylsilane
Synonyms: chloro(chloromethyl)dimethylsilane
Molecular Weight (g/mol): 143.08
Molecular Formula: C3H8Cl2Si
InChi Key: ITKVLPYNJQOCPW-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results