missing translation for 'onlineSavingsMsg'
Learn More

Chloro(chloromethyl)dimethylsilane, 98%, AcroSeal™

Product Code. 11965931 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11965931

Brand: Thermo Scientific Acros 434021000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1719-57-9 | C3H8Cl2Si | 143.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1719-57-9
Color Colorless to Straw
Density 1.0800g/mL
Boiling Point 114.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C3H8Cl2Si
Linear Formula ClCH2Si(CH3)2Cl
Refractive Index 1.4360 to 1.4380
MDL Number MFCD00000875
Quantity 100 mL
Specific Gravity 1.08
Synonym chloro chloromethyl dimethylsilane, chloromethyl dimethylchlorosilane, chloromethyldimethylchlorosilane, silane, chloro chloromethyl dimethyl, cmdmcs, chloro cholromethyl dimethylsilane, ch3 2sicl ch2cl, chloromethylchlorodimethylsilane, dimethylchloromethylchlorosilane, chloro chloromethyl-dimethylsilane
Solubility Information Solubility in water: reacts
InChI Key ITKVLPYNJQOCPW-UHFFFAOYSA-N
SMILES C[Si](C)(Cl)CCl
IUPAC Name chloro-(chloromethyl)-dimethylsilane
Molecular Weight (g/mol) 143.08
PubChem CID 74393
Formula Weight 143.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloro(chloromethyl)dimethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.