CAS RN 17216-62-5
IUPAC Name:
1,3-diethyl 2-(2-cyanoethyl)propanedioate
Synonyms:
1,3-diethyl 2-(2-cyanoethyl)propanedioate
Molecular Weight (g/mol):
213.23
Molecular Formula:
C10H15NO4
InChi Key:
YJJLOESDBPRZIP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(CCC#N)C(=O)OCC
1
–
1
of
1
results
1
Diethyl 2-(2-cyanoethyl)malonate, 98+%
CAS: 17216-62-5 Molecular Formula: C10H15NO4 Molecular Weight (g/mol): 213.233 MDL Number: MFCD00001966 InChI Key: YJJLOESDBPRZIP-UHFFFAOYSA-N PubChem CID: 87005 IUPAC Name: diethyl 2-(2-cyanoethyl)propanedioate SMILES: CCOC(=O)C(CCC#N)C(=O)OCC