missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-(2-cyanoethyl)malonate, 98+%

Product Code. 11434623
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434623 50 g 50g
11424623 10 g 10g
11314509 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434623 Supplier Thermo Scientific Alfa Aesar Supplier No. A11567.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17216-62-5 | C10H15NO4 | 213.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17216-62-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.233
MDL Number MFCD00001966
InChI Key YJJLOESDBPRZIP-UHFFFAOYSA-N
PubChem CID 87005
IUPAC Name diethyl 2-(2-cyanoethyl)propanedioate
SMILES CCOC(=O)C(CCC#N)C(=O)OCC

Specifications

Density 1.079
Boiling Point 115°C to 117°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.436
Quantity 50 g
UN Number UN3276
Beilstein 1790237
Synonym diethyl 2-2-cyanoethyl malonate, diethyl 2-cyanoethyl malonate, 1,3-diethyl 2-2-cyanoethyl propanedioate, diethyl2-2-cyanoethyl malonate, diethyl 2-2-cyanoethyl-malonate, propanedioic acid, 2-cyanoethyl-, diethyl ester, 2-cyanoethylmalonic acid diethylester, diethyl 2-2-cyanoethyl propane-1,3-dioate, pubchem14162
Formula Weight 213.23
Percent Purity ≥98%
Chemical Name or Material Diethyl 2- (2-cyanoethyl)malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.