accessibility menu, dialog, popup

UserName

CAS RN 17302-46-4

CH3OOONOO

CAS RN 17302-46-4

IUPAC Name: 2-(methoxycarbonyl)-4-nitrobenzen-1-olate
Synonyms: 2-(methoxycarbonyl)-4-nitrobenzenolate
Molecular Weight (g/mol): 196.14
Molecular Formula: C8H6NO5
InChi Key: UUBFELFUKFJSRD-UHFFFAOYSA-M
SMILES: COC(=O)C1=CC(=CC=C1[O-])[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results