accessibility menu, dialog, popup

UserName

CAS RN 1731-86-8

IUPAC Name: methyl undecanoate
Synonyms: undecanoic acid, methyl ester
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: XPQPWPZFBULGKT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results