missing translation for 'onlineSavingsMsg'
Learn More

Methyl undecanoate, 97%

Product Code. 10421484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421484 5 g Glass bottle 5g
10133204 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421484 Supplier Thermo Scientific Acros Supplier No. 426580050
 Restricted Item
Estimated Shipment: 21-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1731-86-8 | C12H24O2 | 200.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-86-8
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00008957
InChI Key XPQPWPZFBULGKT-UHFFFAOYSA-N
Synonym undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
PubChem CID 15607
ChEBI CHEBI:87527
IUPAC Name methyl undecanoate
SMILES CCCCCCCCCCC(=O)OC

Specifications

Color Colorless to Yellow
Density 0.8720g/mL
Boiling Point 246°C to 248°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9CO2CH3
Refractive Index 1.429
Quantity 5 g
Specific Gravity 0.872
Solubility Information Solubility in water: insoluble
Formula Weight 200.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl undecanoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl undecanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.