accessibility menu, dialog, popup

UserName

OON

CAS RN 173282-69-4

IUPAC Name: 6-phenoxypyridine-3-carbaldehyde
Synonyms: 6-phenoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: SOPOQXNWJNVZMI-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OC2=CC=CC=C2)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results