missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxynicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10774755
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10774755 5 g Amber Glass Bottle 5g
10551512 250 mg Amber Glass Bottle 250mg
10487032 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10774755 Supplier Thermo Scientific Maybridge Supplier No. CC19604DE
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 173282-69-4
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD02681950
InChI Key SOPOQXNWJNVZMI-UHFFFAOYSA-N
PubChem CID 2776500
IUPAC Name 6-phenoxypyridine-3-carbaldehyde
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 6-Phenoxynicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.