accessibility menu, dialog, popup

UserName

NH2SHNFFF

CAS RN 1736-70-5

IUPAC Name: [3-(trifluoromethyl)phenyl]thiourea
Synonyms: 3-(trifluoromethyl)phenylthiourea
Molecular Weight (g/mol): 220.21
Molecular Formula: C8H7F3N2S
InChi Key: WKRUQAYFMKZMPJ-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results