accessibility menu, dialog, popup

UserName

ONOFN

CAS RN 17417-09-3

IUPAC Name: 2-fluoro-5-nitrobenzonitrile
Synonyms: 2-fluoro-5-nitrobenzonitrile
Molecular Weight (g/mol): 166.11
Molecular Formula: C7H3FN2O2
InChi Key: YLACBMHBZVYOAP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(=C1)C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)