missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 10622273
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10622273 10 g 10g
10385152 1 g 1g
10481124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10622273 Supplier Thermo Scientific Maybridge Supplier No. TL00351EA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17417-09-3
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.11
MDL Number MFCD00042299
InChI Key YLACBMHBZVYOAP-UHFFFAOYSA-N
Synonym benzonitrile, 2-fluoro-5-nitro, 3-cyano-4-fluoronitrobenzene, 2-fluoro-5-nitro-benzonitrile, 2-fluoro-5-nitrobenzenecarbonitrile, 2-fluoro-5-nitrobenzenenitrile, 3-cyano-4-fluoro-1-nitrobenzene, pubchem4792, intermediates-zcf02297, acmc-209e7w, 5-nitro-2-fluorobenzonitrile
PubChem CID 519417
IUPAC Name 2-fluoro-5-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(F)C(=C1)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 166.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.