accessibility menu, dialog, popup

UserName

CAS RN 175137-65-2

IUPAC Name: 4-(4-chlorobenzenesulfonyl)-3-methylthiophene-2-carboxylic acid
Synonyms: 4-(4-chlorobenzenesulfonyl)-3-methylthiophene-2-carboxylic acid
Molecular Weight (g/mol): 316.77
Molecular Formula: C12H9ClO4S2
InChi Key: WZQQSJVPCVCQKE-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1S(=O)(=O)C1=CC=C(Cl)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results