accessibility menu, dialog, popup

UserName

FSOONOHNH2(E/Z)

CAS RN 175203-76-6

IUPAC Name: 2-(4-fluorobenzenesulfonyl)-N'-hydroxyethanimidamide
Synonyms: 2-(4-fluorobenzenesulfonyl)-N'-hydroxyethanimidamide
Molecular Weight (g/mol): 232.23
Molecular Formula: C8H9FN2O3S
InChi Key: STPCRDRACVTHTE-UHFFFAOYSA-N
SMILES: NC(CS(=O)(=O)C1=CC=C(F)C=C1)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results