accessibility menu, dialog, popup

UserName

FON

CAS RN 175204-06-5

IUPAC Name: 2-fluoro-6-phenoxybenzonitrile
Synonyms: 2-fluoro-6-phenoxybenzonitrile
Molecular Weight (g/mol): 213.21
Molecular Formula: C13H8FNO
InChi Key: FBHXELGUZCMKQY-UHFFFAOYSA-N
SMILES: FC1=CC=CC(OC2=CC=CC=C2)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results