accessibility menu, dialog, popup

UserName

CH3OOHNCH3ONOO

CAS RN 175204-17-8

IUPAC Name: ethyl 4-acetamido-3-nitrobenzoate
Synonyms: ethyl 4-acetamido-3-nitrobenzoate
Molecular Weight (g/mol): 252.23
Molecular Formula: C11H12N2O5
InChi Key: AQEATNDLNBFMRO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(NC(C)=O)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results