accessibility menu, dialog, popup

UserName

CAS RN 175204-40-7

CH3H3CCH3NONCl

CAS RN 175204-40-7

IUPAC Name: 3-(4-tert-butylphenyl)-5-(chloromethyl)-1,2,4-oxadiazole
Synonyms: 3-(4-tert-butylphenyl)-5-(chloromethyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 250.73
Molecular Formula: C13H15ClN2O
InChi Key: ZLQQUIHBQATFOP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=NOC(CCl)=N1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results