accessibility menu, dialog, popup

UserName

NCSOFFF

CAS RN 175205-33-1

IUPAC Name: 1-isothiocyanato-2-(trifluoromethoxy)benzene
Synonyms: 1-isothiocyanato-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 219.18
Molecular Formula: C8H4F3NOS
InChi Key: ZFEJYPIWOGCCFS-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results