missing translation for 'onlineSavingsMsg'
Learn More

2-(trifluoromethoxy)phenyl isothiocyanate, Thermo Scientific™

Product Code. 10285594
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10285594 25 g 25g
10119382 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285594 Supplier Thermo Scientific Maybridge Supplier No. SB01674EB
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-33-1
Molecular Formula C8H4F3NOS
Molecular Weight (g/mol) 219.18
MDL Number MFCD00220860
InChI Key ZFEJYPIWOGCCFS-UHFFFAOYSA-N
PubChem CID 2777336
IUPAC Name 1-isothiocyanato-2-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC=C1N=C=S

Specifications

Density 1.3540g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Synonym 2-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethoxy benzene, benzene,1-isothiocyanato-2-trifluoromethoxy, 1-isothiocyanato-2-trifluoromethoxy-benzene, 2-trifluoromethoxy phenylisothiocyanate, benzene, 1-isothiocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisothiocyanate, 1-isothiocyanato-2-trifluoromethyloxy benzene, 2-isothiocyanato-alpha,alpha,alpha-trifluoroanisole
Formula Weight 219.19
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.