accessibility menu, dialog, popup

UserName

FNCSBr

CAS RN 175205-35-3

IUPAC Name: 2-bromo-4-fluoro-1-isothiocyanatobenzene
Synonyms: 2-bromo-4-fluoro-1-isothiocyanatobenzene
Molecular Weight (g/mol): 232.07
Molecular Formula: C7H3BrFNS
InChi Key: SSLVXFSUADEZBQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(N=C=S)C(Br)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results