accessibility menu, dialog, popup

UserName

CAS RN 175205-89-7

IUPAC Name: 2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 335.93
Molecular Formula: C9H4BrCl2F3O
InChi Key: MSMAFPRJZLLQNJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C(=O)CBr)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results