missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethan-1-one, Thermo Scientific™

Product Code. 10143234
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143234 5 g Amber Glass Bottle 5g
10059471 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143234 Supplier Thermo Scientific Maybridge Supplier No. SEW04510DE
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-89-7
Molecular Formula C9H4BrCl2F3O
Molecular Weight (g/mol) 335.93
MDL Number MFCD00221016
InChI Key MSMAFPRJZLLQNJ-UHFFFAOYSA-N
Synonym 2-bromo-2',6'-dichloro-4'-trifluoromethyl-acetophenone, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethanone, ethanone,2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl, 2-bromo-2,6-dichloro-4-trifluoromethyl-acetophenone, 2-bromo-2',6'-dichloro-4'-trifluoromethyl acetophenone, 1-2,6-bis chloranyl-4-trifluoromethyl phenyl-2-bromanyl-ethanone
PubChem CID 2781609
IUPAC Name 2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethanone
SMILES FC(F)(F)C1=CC(Cl)=C(C(=O)CBr)C(Cl)=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 335.93
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[2, 6-dichloro-4-(trifluoromethyl)phenyl]ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.