accessibility menu, dialog, popup

UserName

CAS RN 175277-03-9

CH3OONSFFFCH3

CAS RN 175277-03-9

IUPAC Name: ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 315.31
Molecular Formula: C14H12F3NO2S
InChi Key: LPIXRQSYBTUXOQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=CC=C(C=C1)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)