accessibility menu, dialog, popup

UserName

CH3NNOHOCH3H3CCH3

CAS RN 175277-09-5

IUPAC Name: 1-tert-butyl-3-methyl-1H-pyrazole-5-carboxylic acid
Synonyms: 2-tert-butyl-5-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 182.22
Molecular Formula: C9H14N2O2
InChi Key: JZPMLZWJUMATOQ-UHFFFAOYSA-N
SMILES: CC1=NN(C(=C1)C(O)=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results