missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10586995
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10586995 5 g 5g
10008171 1 g 1g
10485142 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10586995 Supplier Thermo Scientific Maybridge Supplier No. SPB03128DE
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-09-5
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.223
MDL Number MFCD00084917
InChI Key JZPMLZWJUMATOQ-UHFFFAOYSA-N
Synonym 1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic acid, 2-tert-butyl-5-methyl-pyrazole-3-carboxylic acid, 1-tert-butyl-3-methylpyrazole-5-carboxylic acid, maybridge1_008627, buttpark 9218-70, 2-tert-butyl-5-methyl-3-pyrazolecarboxylic acid, 1-tert-butyl-5-carboxy-3-methyl-1h-pyrazole, 1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic acid
PubChem CID 2744573
IUPAC Name 2-tert-butyl-5-methylpyrazole-3-carboxylic acid
SMILES CC1=NN(C(=C1)C(=O)O)C(C)(C)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.