accessibility menu, dialog, popup

UserName

NClNH2FFF

CAS RN 175277-74-4

IUPAC Name: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine
Synonyms: 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methanamine
Molecular Weight (g/mol): 210.58
Molecular Formula: C7H6ClF3N2
InChi Key: JVQYWHGODHSTAM-UHFFFAOYSA-N
SMILES: NCC1=C(Cl)C=C(C=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results