accessibility menu, dialog, popup

UserName

OHOOFFFNH2

CAS RN 175278-22-5

IUPAC Name: 4-amino-3-(trifluoromethoxy)benzoic acid
Synonyms: 4-amino-3-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 221.14
Molecular Formula: C8H6F3NO3
InChi Key: IXJFWBNYFTWBOR-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=C(C=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results