accessibility menu, dialog, popup

UserName

H3CBrSOOCl

CAS RN 175278-24-7

IUPAC Name: 4-bromo-2-ethylbenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-ethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 283.56
Molecular Formula: C8H8BrClO2S
InChi Key: UOIVESXBURLTPX-UHFFFAOYSA-N
SMILES: CCC1=C(C=CC(Br)=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results