missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-ethylbenzene-1-sulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10621893
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10621893 1 g Amber Glass Bottle 1g
10395152 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10621893 Supplier Thermo Scientific Maybridge Supplier No. TL00690DA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-24-7
Molecular Formula C8H8BrClO2S
Molecular Weight (g/mol) 283.564
MDL Number MFCD00204178
InChI Key UOIVESXBURLTPX-UHFFFAOYSA-N
Synonym 4-bromo-2-ethylbenzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulfonylchloride, 4-bromo-2-ethylbenzenesulphonyl chloride, benzenesulfonylchloride, 4-bromo-2-ethyl, pubchem5062, 4-bromo-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethylphenylsulfonyl chloride, 4-bromanyl-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethyl-benzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulphonyl chloride
PubChem CID 2737536
IUPAC Name 4-bromo-2-ethylbenzenesulfonyl chloride
SMILES CCC1=C(C=CC(=C1)Br)S(=O)(=O)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 283.57
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-ethylbenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.