accessibility menu, dialog, popup

UserName

CH3NNNH2NCH3H2O

CAS RN 175278-59-8

IUPAC Name: dimethyl-1,3,5-triazin-2-amine hydrate
Synonyms: dimethyl-1,3,5-triazin-2-amine hydrate
Molecular Weight (g/mol): 142.16
Molecular Formula: C5H10N4O
InChi Key: CZKUOCRCKNMNTI-UHFFFAOYSA-N
SMILES: O.CC1=NC(N)=NC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results