missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-1,3,5-triazin-2-amine hydrate, ≥95%, Thermo Scientific™

Product Code. 10772873
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772873 10 g Amber Glass Bottle 10g
10334392 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772873 Supplier Thermo Scientific Maybridge Supplier No. RF02292EA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-59-8
Molecular Formula C5H10N4O
Molecular Weight (g/mol) 142.16
MDL Number MFCD00663061
InChI Key CZKUOCRCKNMNTI-UHFFFAOYSA-N
Synonym 4,6-dimethyl-1,3,5-triazin-2-amine hydrate, dimethyl-1,3,5-triazin-2-amine hydrate, 1,3,5-triazin-2-amine,4,6-dimethyl-, hydrate 1:1, 2-amino-4,6-dimethyl-1,3,5-triazine hydrate, 4,6-dimethyl-1,3,5-triazine-2-ylamine, oxamethane, 1,3,5-triazin-2-amine,4,6-dimethyl-,hydrate 1:1
PubChem CID 2782054
IUPAC Name 4,6-dimethyl-1,3,5-triazin-2-amine;hydrate
SMILES O.CC1=NC(N)=NC(C)=N1

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 142.17
Percent Purity 95%
Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.