accessibility menu, dialog, popup

UserName

NH2ONOOSOOO

CAS RN 175278-60-1

IUPAC Name: 2-amino-6-nitro-4-sulfonatobenzen-1-olate
Synonyms: 2-amino-6-nitro-4-sulfonatobenzenolate
Molecular Weight (g/mol): 232.17
Molecular Formula: C6H4N2O6S
InChi Key: RHPXYZMDLOJTFF-UHFFFAOYSA-L
SMILES: NC1=CC(=CC(=C1[O-])[N+]([O-])=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results