accessibility menu, dialog, popup

UserName

NNFFF

CAS RN 17573-78-3

IUPAC Name: 4,5,6-trifluoropyrimidine
Synonyms: 4,5,6-trifluoropyrimidine
Molecular Weight (g/mol): 134.06
Molecular Formula: C4HF3N2
InChi Key: AQKOJKCYBNUFLU-UHFFFAOYSA-N
SMILES: FC1=NC=NC(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results