missing translation for 'onlineSavingsMsg'
Learn More

4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals

Product Code. 11415600
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415600 1 g 1g
15403327 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415600 Supplier Thermo Scientific Alfa Aesar Supplier No. H25855.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17573-78-3 | Molecular Formula: C4HF3N2 | Molecular Weight: 134.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17573-78-3
Molecular Formula C4HF3N2
Molecular Weight (g/mol) 134.06
MDL Number MFCD05662363
InChI Key AQKOJKCYBNUFLU-UHFFFAOYSA-N
Synonym pyrimidine, 4,5,6-trifluoro, pubchem6922, acmc-1bol2, pyrimidine,4,5,6-trifluoro, 4,5,6-trifluoro-1,3-diazine, 4,5,6-tris fluoranyl pyrimidine, 4,5,6-trifluoropyrimidine, 4,5,6-trifluoro pyrimidine, pyrimidine, 4,5,6-trifluoro-8ci,9ci
PubChem CID 3572764
IUPAC Name 4,5,6-trifluoropyrimidine
SMILES FC1=NC=NC(F)=C1F

Specifications

Density 1.4609
Boiling Point 83°C to 85°C
Flash Point >85°C (185°F)
Refractive Index 1.408
Quantity 1 g
UN Number UN1760
Formula Weight 134.06
Percent Purity 97%
Chemical Name or Material 4,5,6-Trifluoropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.