accessibility menu, dialog, popup

UserName

CAS RN 1758-54-9

H3CNONH(Z)

CAS RN 1758-54-9

IUPAC Name: (2Z)-2-(1-nitrosoethylidene)-1,2-dihydropyridine
Synonyms: (2Z)-2-(1-nitrosoethylidene)-1H-pyridine
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: KWYHBFKEFFMBHW-SREVYHEPSA-N
SMILES: C\C(N=O)=C1\NC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results