missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-pyridyl ketoxime, 97%

Product Code. 15498827
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15498827

Brand: Thermo Scientific Alfa Aesar H52232.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1758-54-9 | C7H8N2O | 136.15 g/mol

Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-54-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00605540
InChI Key KWYHBFKEFFMBHW-SREVYHEPSA-N
Synonym 1-pyridin-2-yl ethanone oxime, 1-2-pyridinyl-1-ethanone oxime, 1-2-pyridinyl ethanone oxime, 1-pyridin-2-yl-ethanone oxime, 1-pyridin-2-ylethan-1-one oxime, e-1-pyridin-2-yl ethanone oxime, e-n-1-pyridin-2-yl ethylidene hydroxylamine, methyl2-pyridylketoneoxime, 2-1-nitrosoethylidene-1h-pyridine, e-1-pyridine-2-yl ethanone oxime
PubChem CID 6740780
SMILES C\C(N=O)=C1\NC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name 2-(1-nitrosoethylidene)-1H-pyridine
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Methyl 2-pyridyl ketoxime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.