accessibility menu, dialog, popup

UserName

CAS RN 1760-24-3

IUPAC Name: hydrogen (2-aminoethyl)[3-(trimethoxysilyl)propyl]amine chloride
Synonyms: hydrogen (2-aminoethyl)[3-(trimethoxysilyl)propyl]amine chloride
Molecular Weight (g/mol): 258.82
Molecular Formula: C8H23ClN2O3Si
InChi Key: SZBIXNNLIKDOEJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CO[Si](CCCNCCN)(OC)OC
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results