missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%

Product Code. 11488816
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488816 250 g 250g
11478816 50 g 50g
11468816 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488816 Supplier Thermo Scientific Alfa Aesar Supplier No. A16981.30
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1760-24-3 | C8H22N2O3Si | 222.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1760-24-3
Molecular Formula C8H22N2O3Si
Molecular Weight (g/mol) 222.36
MDL Number MFCD00008173
InChI Key PHQOGHDTIVQXHL-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
PubChem CID 15659
IUPAC Name N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
SMILES CO[Si](CCCNCCN)(OC)OC

Specifications

Boiling Point 261°C to 263°C
Quantity 250 g
Flash Point 136°C (276°F)
Refractive Index 1.442
Sensitivity Air and moisture sensitive
Density 1.01
UN Number UN2735
Beilstein 636230
Formula Weight 222.36
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(2-Aminoethylamino)propyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.